Learn More
Chemglass Life Sciences Thermocouple Clamp, PP
SDP
Catalog No. NC1390128
Encompass_Preferred
Thermocouple Clamp, Polypropylene. Provides the eastiest and most secure way to hold thermocouple probes in the laboratory. Accepts 1/16", 1/8" and 1/4" diameter probes. The thermocouple probe is inserted through the opening at the bottom of the clamp and the thumbscrew is tightened against the plug. Stainless steel support arm is 1/4" diameter x 8" long.
Catalog No. NC1390128 Supplier Chemglass Life Sciences Supplier No. CG3499H10
Thermocouple Clamp, Polypropylene. Provides the eastiest and most secure way to hold thermocouple probes in the laboratory. Accepts 1/16", 1/8" and 1/4" diameter probes. The thermocouple probe is inserted through the opening at the bottom of the clamp and the thumbscrew is tightened against the plug. Stainless steel support arm is 1/4" diameter x 8" long.