Learn More
Chemglass Life Sciences Clamp, Thermometer, 8.5" OAL
SDP
Catalog No. 501311237
Encompass_Preferred
Clamp, Thermometer, 8.5" OAL. Clamp will hold thermometers and other small diameter items from 1/4 to 1/2¿ (6.5 to 12.7mm). Overall length is 8.5¿ (220mm).
Catalog No. 50-131-1237 Supplier Chemglass Life Sciences Supplier No. CG920110
Clamp, Thermometer, 8.5" OAL. Clamp will hold thermometers and other small diameter items from 1/4 to 1/2¿ (6.5 to 12.7mm). Overall length is 8.5¿ (220mm).