Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Clamp Ties with Curved Head, White Plastic; 100/PK
Catalog No. NC9616967
Encompass
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".
Catalog No. NC9616967 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9561500
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".