Learn More
Chemglass Life Sciences Nylon Tubing Clamp, Clamping Range: 1.406-1.61in, 35.71-40.89mm
SDP
Catalog No. 501311055
Encompass_Preferred
Nylon Tubing Clamp, Clamping Range: 1.406-1.61in, 35.71-40.89mm• Ideal for vacuum, pressure and liquid applications• Chemical- and corrosion-resistant• Can be sterilized for biological and medical applications• Easy to install and remove by hand or with pliers• Push down on one side to release
Catalog No. 50-131-1055 Supplier Chemglass Life Sciences Supplier No. CG15222
Nylon Tubing Clamp, Clamping Range: 1.406-1.61in, 35.71-40.89mm• Ideal for vacuum, pressure and liquid applications• Chemical- and corrosion-resistant• Can be sterilized for biological and medical applications• Easy to install and remove by hand or with pliers• Push down on one side to release