Learn More
Kuhner Shaker Inc CLAMP TUBESPIN 600ML BIOREACTO
Catalog No. 502570292
Encompass
Clamp TubeSpin 600mL Bioreactor - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 600ml TPP TubeSpin Bioreactors
Catalog No. 50-257-0292 Supplier Kuhner Shaker Inc Supplier No. SM310600
Clamp TubeSpin 600mL Bioreactor - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 600ml TPP TubeSpin Bioreactors