Learn More
Hach Company Clamp, Tubing, Large, 12/pk
Supplier:  Hach Company 2087901
Encompass
Molded, white polycarbonate. Autoclavable. Will not distort, rust or corrode. Mini-clamp is a single position on/off clamp for use with thin-walled tubing under 4.7 mm (3/16") O.D. Larger clamp has 15 positions, stepped control to throttle flow from full open to complete closure. Fits tubing from 3.2 mm (1/8") to 11 mm (3/8") O.D.
Catalog No. 50-199-5289