Learn More
Kuhner Shaker Inc CLAMP U-1000 - SINGLE PLASTIC
Catalog No. 502570291
Encompass
Clamp U-1000 - Single plastic clamp for use with Kuhner Shaker Universal Trays. Ideal for use with Kuhner TOM Online CO2 and O2 Transfer Rate Monitoring System. Holds 1000ml flasks
Catalog No. 50-257-0291 Supplier Kuhner Shaker Inc Supplier No. SM311000K
Clamp U-1000 - Single plastic clamp for use with Kuhner Shaker Universal Trays. Ideal for use with Kuhner TOM Online CO2 and O2 Transfer Rate Monitoring System. Holds 1000ml flasks