Learn More
Kuhner Shaker Inc CLAMP U-150 - SINGLE REMOVABLE
Catalog No. 502570297
Encompass
Clamp U-150 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 150ml flasks
Catalog No. 50-257-0297 Supplier Kuhner Shaker Inc Supplier No. SM310150
Clamp U-150 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 150ml flasks