Learn More
Kuhner Shaker Inc CLAMP U-1500 - SINGLE REMOVABL
Catalog No. 502570290
Encompass
Clamp U-1500 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 1500ml flasks
Catalog No. 50-257-0290 Supplier Kuhner Shaker Inc Supplier No. SM311500
Clamp U-1500 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 1500ml flasks