Learn More
Kuhner Shaker Inc CLAMP U-1600 THOMSON - SINGLE
Catalog No. 502570289
Encompass
Clamp U-1600 Thomson - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 1600ml Thomson flasks
Catalog No. 50-257-0289 Supplier Kuhner Shaker Inc Supplier No. SM311600
Clamp U-1600 Thomson - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 1600ml Thomson flasks