Learn More
Kuhner Shaker Inc CLAMP U-200 - SINGLE REMOVABLE
Catalog No. 502570296
Encompass
Clamp U-200 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 200ml flasks
Catalog No. 50-257-0296 Supplier Kuhner Shaker Inc Supplier No. SM310200
Clamp U-200 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 200ml flasks