Learn More
Kuhner Shaker Inc CLAMP U-250 - SINGLE PLASTIC C
Catalog No. 502570295
Encompass
Clamp U-250 - Single plastic clamp for use with Kuhner Shaker Universal Trays. Ideal for use with Kuhner TOM Online CO2 and O2 Transfer Rate Monitoring System. Holds 250ml flasks
Catalog No. 50-257-0295 Supplier Kuhner Shaker Inc Supplier No. SM310250K
Clamp U-250 - Single plastic clamp for use with Kuhner Shaker Universal Trays. Ideal for use with Kuhner TOM Online CO2 and O2 Transfer Rate Monitoring System. Holds 250ml flasks