Learn More
Kuhner Shaker Inc CLAMP U-2800 FERNBACH - SINGLE
Catalog No. 502570288
Encompass
Clamp U-2800 Fernbach - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 2800ml Fernbach flasks
Catalog No. 50-257-0288 Supplier Kuhner Shaker Inc Supplier No. SM312800F
Clamp U-2800 Fernbach - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 2800ml Fernbach flasks