Learn More
Kuhner Shaker Inc CLAMP U-300 - SINGLE REMOVABLE
Catalog No. 502570294
Encompass
Clamp U-300 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 300ml flasks
Catalog No. 50-257-0294 Supplier Kuhner Shaker Inc Supplier No. SM310300
Clamp U-300 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 300ml flasks