Learn More
Kuhner Shaker Inc CLAMP U-3000 - SINGLE REMOVABL
Catalog No. 502570287
Encompass
Clamp U-3000 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 3000ml flasks
Catalog No. 50-257-0287 Supplier Kuhner Shaker Inc Supplier No. SM313000
Clamp U-3000 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 3000ml flasks