Learn More
Kuhner Shaker Inc CLAMP U-4000 - SINGLE REMOVABL
Catalog No. 502570286
Encompass
Clamp U-4000 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 4000ml flasks
Catalog No. 50-257-0286 Supplier Kuhner Shaker Inc Supplier No. SM314000
Clamp U-4000 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 4000ml flasks