Learn More
Kuhner Shaker Inc CLAMP U-500 - FOR ERLENMEYER
Catalog No. NC1751812
Encompass
Clamp U-500 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 500ml flasks
Catalog No. NC1751812 Supplier Kuhner Shaker Inc Supplier No. SM310500
Clamp U-500 - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 500ml flasks