Learn More
Kuhner Shaker Inc CLAMP U-500 - SINGLE PLASTIC C
Catalog No. 502570293
Encompass
Clamp U-500 - Single plastic clamp for use with Kuhner Shaker Universal Trays. Ideal for use with Kuhner TOM Online CO2 and O2 Transfer Rate Monitoring System. Holds 500ml flasks
Catalog No. 50-257-0293 Supplier Kuhner Shaker Inc Supplier No. SM310500K
Clamp U-500 - Single plastic clamp for use with Kuhner Shaker Universal Trays. Ideal for use with Kuhner TOM Online CO2 and O2 Transfer Rate Monitoring System. Holds 500ml flasks