Learn More
Electron Microscopy Sciences Tripod Polisher Clamping "L" Bracket
SDP
Catalog No. 50980869
Encompass
Used in same way as Screw Clamp L-Bracket with different method of clamping. Inserts are held tightly with good thermal contact for mounting samples when the Lbracket is heated on a hot plate, Can be lit from the bottom.
Catalog No. 50-980-869 Supplier Electron Microscopy Sciences Supplier No. 5011630
Used in same way as Screw Clamp L-Bracket with different method of clamping. Inserts are held tightly with good thermal contact for mounting samples when the Lbracket is heated on a hot plate, Can be lit from the bottom.