Learn More
Leica Biosystems Clamping Lever, Long - Replacement for Leica CM1850, CM1510 S, and 21 Series Microtomes
Catalog No. NC3872775
Encompass
Genuine OEM replacement long clamping lever designed to restore secure clamping and precise adjustment on compatible microtomes and cryostats. It is intended for use during maintenance and repair to replace worn or damaged levers and maintain instrument performance.
  • Provides secure clamping for specimen and component adjustment.
  • Long lever profile improves reach and ergonomics.
  • Simple installation for maintenance and repairs.
  • Restores original clamping function and repeatability.
Catalog No. NC3872775 Supplier Leica Biosystems Supplier No. 14050231715
Genuine OEM replacement long clamping lever designed to restore secure clamping and precise adjustment on compatible microtomes and cryostats. It is intended for use during maintenance and repair to replace worn or damaged levers and maintain instrument performance.
  • Provides secure clamping for specimen and component adjustment.
  • Long lever profile improves reach and ergonomics.
  • Simple installation for maintenance and repairs.
  • Restores original clamping function and repeatability.