Learn More
Fritsch Milling And Sizing Inc Clamping Lid 100mm Cpl. with Security Glass
SDP
Catalog No. 501534182
Encompass
Clamping lid POM plastic with security glass for dry sieving with test sieves 100 mm dia. and grinding head for conversion of the ANALYSETTE 3 to the PULVERISETTE 0.
The clamping lid with security glass allows the operator to observe the sieving process and the ANALYSETTE 3 can be used with this clamping lid as Vibratory Micro Mill PULVERISETTE 0 for milling and homogenizing of small quantities of samples (sample quantity: 0.1 – 10 ml, max. feed size: < 5 mm, final fineness: 5 – 10 µm).
Catalog No. 50-153-4182 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.2016.00
Clamping lid POM plastic with security glass for dry sieving with test sieves 100 mm dia. and grinding head for conversion of the ANALYSETTE 3 to the PULVERISETTE 0.
The clamping lid with security glass allows the operator to observe the sieving process and the ANALYSETTE 3 can be used with this clamping lid as Vibratory Micro Mill PULVERISETTE 0 for milling and homogenizing of small quantities of samples (sample quantity: 0.1 – 10 ml, max. feed size: < 5 mm, final fineness: 5 – 10 µm).