Learn More
Fritsch Milling And Sizing Inc Clamping Lid 200Mm/8"
SDP
Catalog No. 501534183
Encompass
For dry sieving with test sieves 200 mm/8" dia.
The clamping lid made of plexiglas allows the operator to observe the sieving process.
For applications, where it is not required to observe the sieving process, a clamping lid without window insert, article number 31.2050.00 is also available.
This clamping lid can also be used if you would like to use the Vibratory Micro Mill PULVERISETTE 0 as Vibratory Sieve Shaker ANALYSETTE 3 SPARTAN. The needed sieve pan and the sieves must be ordered separately.
Catalog No. 50-153-4183 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.2020.00
For dry sieving with test sieves 200 mm/8" dia.
The clamping lid made of plexiglas allows the operator to observe the sieving process.
For applications, where it is not required to observe the sieving process, a clamping lid without window insert, article number 31.2050.00 is also available.
This clamping lid can also be used if you would like to use the Vibratory Micro Mill PULVERISETTE 0 as Vibratory Sieve Shaker ANALYSETTE 3 SPARTAN. The needed sieve pan and the sieves must be ordered separately.