Learn More
Fritsch Milling And Sizing Inc Clamping Lid 100 mm with 1 Nozzle
SDP
Catalog No. 501534184
Encompass
With 1 nozzles for wet sieving with test sieves 100 mm dia.
The wet clamping lid features 1 nozzle which sprays the entire interior chamber, thus helping to flush the material through the sieve apertures.
Especially recommended for materials with a high electrostatic charge or very high content of fine material.
Catalog No. 50-153-4184 Supplier Fritsch Milling And Sizing Inc Supplier No. 31.2040.00
Add to Cart
Add to Cart
With 1 nozzles for wet sieving with test sieves 100 mm dia.
The wet clamping lid features 1 nozzle which sprays the entire interior chamber, thus helping to flush the material through the sieve apertures.
Especially recommended for materials with a high electrostatic charge or very high content of fine material.