Learn More
Chemglass Life Sciences Clamp, Nester
SDP
Catalog No. CG923101
Encompass_Preferred
Clamp, Nester. Strong brass chromed chain and band with stainless steel rod for holding distillation columns, odd-shaped glassware, etc. Clamp holds apparatus with diameters from 38mm to 102mm.
Catalog No. CG923101 Supplier Chemglass Life Sciences Supplier No. CG923101
Clamp, Nester. Strong brass chromed chain and band with stainless steel rod for holding distillation columns, odd-shaped glassware, etc. Clamp holds apparatus with diameters from 38mm to 102mm.