Learn More
Kuhner Shaker Inc CLAMPS FOR GROWTH FLASKS
Catalog No. NC1980814
Encompass
Clamp U-2800 Thomson - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 2800ml Thomson flasks
Catalog No. NC1980814 Supplier Kuhner Shaker Inc Supplier No. SM312800T
Clamp U-2800 Thomson - Single removable stainless steel spring-clamp for use with Kuhner Shaker Universal Trays. Holds 2800ml Thomson flasks