Learn More
Med Vet International Lorna-Edna Towel Forceps, 5.25in, Non-Perforating Teeth, German Grade, Von Klaus
Catalog No. 502091718
Encompass_Preferred
5.25in. Lorna Towel Forceps is a German grade surgical instrument. Lorna Towel Forceps have a non perforating clamp that is used to secure instruments and surgical material. Lorna Towel Clamps can also be known as a Edna Towel Clamp. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1718 Supplier Med Vet International Supplier No. VK1175714
5.25in. Lorna Towel Forceps is a German grade surgical instrument. Lorna Towel Forceps have a non perforating clamp that is used to secure instruments and surgical material. Lorna Towel Clamps can also be known as a Edna Towel Clamp. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.