Learn More
Electron Microscopy Sciences Transparent Clamshell Wafer Container, 6", Polycarbonate Material
SDP
Catalog No. 501929911
Encompass_Preferred
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring.
Use: 6" (150mm)
Material: Polycarbonate
Catalog No. 50-192-9911 Supplier Electron Microscopy Sciences Supplier No. 649206
The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring.
Use: 6" (150mm)
Material: Polycarbonate