Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Claramine hydrochloride | 1430194-56-1 | 99.1% | 625.45 g/mol | C37H73ClN4O | 10 MG
SDP

Catalog No. 5000285540
Encompass_Preferred

Claramine hydrochloride is a steroidal polyamine research compound reported to cross the blood-brain barrier and to modulate lipid membrane properties. It is supplied as a powder for laboratory research use only.

  • Molecular formula: C37H73ClN4O.
  • Molecular weight: 625.45 g/mol.
  • Cas number: 1430194-56-1.
  • Form: powder, white to off-white solid.
  • Purity: 99.1% (HPLC, batch certificate of analysis).
  • Solubility: DMSO 2.2 mg/mL (3.52 mM; ultrasonic warming to 60°C recommended).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
  • Intended use: for research use only, not for human or clinical applications.

Catalog No. 50-002-85540 Supplier Medchemexpress LLC Supplier No. HY160791B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Claramine hydrochloride is a steroidal polyamine research compound reported to cross the blood-brain barrier and to modulate lipid membrane properties. It is supplied as a powder for laboratory research use only.

  • Molecular formula: C37H73ClN4O.
  • Molecular weight: 625.45 g/mol.
  • Cas number: 1430194-56-1.
  • Form: powder, white to off-white solid.
  • Purity: 99.1% (HPLC, batch certificate of analysis).
  • Solubility: DMSO 2.2 mg/mL (3.52 mM; ultrasonic warming to 60°C recommended).
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).
  • Intended use: for research use only, not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.