Learn More
Medchemexpress LLC Claramine (hydrochloride) | 99.6% | 625.45 | 5 MG
SDP
Catalog No. 5000394147
Encompass_Preferred
Claramine hydrochloride is a steroid polyamine with blood-brain barrier permeability. It can regulate the properties of lipid membranes and protect cells from various biological toxins, including misfolded protein oligomers and biological protein-based toxins.
  • Steroid polyamine
  • Blood-brain barrier permeability
  • Regulates properties of lipid membranes
  • Protects cells from various biological toxins, including misfolded protein oligomers and biological protein-based toxins
Catalog No. 50-003-94147 Supplier Medchemexpress LLC Supplier No. HY160791B5MG
Claramine hydrochloride is a steroid polyamine with blood-brain barrier permeability. It can regulate the properties of lipid membranes and protect cells from various biological toxins, including misfolded protein oligomers and biological protein-based toxins.
  • Steroid polyamine
  • Blood-brain barrier permeability
  • Regulates properties of lipid membranes
  • Protects cells from various biological toxins, including misfolded protein oligomers and biological protein-based toxins