Learn More
Medchemexpress LLC Claramine TFA | 3030428-57-7 | 95.0% | 703.02 | 5 MG
SDP
Catalog No. 5000394146
Encompass_Preferred
Claramine TFA is a steroidal polyamine that regulates the properties of lipid membranes and offers protection to cells against various biological toxins. These toxins include misfolded protein oligomers and those derived from biological proteins.
  • Does not affect cell viability in human neuroblastoma cells (sh-sy5y) at concentrations below 10 μM.
  • Does not impact cell activity in hek293 cells.
  • Protects human neuroblastoma (sh-sy5y) cells from the harmful effects of pore-forming agents, melittin and α-hemolysin, by inhibiting their binding to cell membranes.
Catalog No. 50-003-94146 Supplier Medchemexpress LLC Supplier No. HY160791A5MG
Claramine TFA is a steroidal polyamine that regulates the properties of lipid membranes and offers protection to cells against various biological toxins. These toxins include misfolded protein oligomers and those derived from biological proteins.
  • Does not affect cell viability in human neuroblastoma cells (sh-sy5y) at concentrations below 10 μM.
  • Does not impact cell activity in hek293 cells.
  • Protects human neuroblastoma (sh-sy5y) cells from the harmful effects of pore-forming agents, melittin and α-hemolysin, by inhibiting their binding to cell membranes.