Learn More
Sigma Aldrich Fine Chemicals Biosciences Claramine trifluoroacetate25MG
Catalog No. 502904558
Encompass
Claramine is an easy and quick to synthesize polyaminosteroid derivative. It is a trodusquemine-related compound and shares with it a spermino group which is responsible for its inhibitory action towards protein tyrosine phosphatase 1B (PTP1B).
Catalog No. 50-290-4558 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SML154525MG
Claramine is an easy and quick to synthesize polyaminosteroid derivative. It is a trodusquemine-related compound and shares with it a spermino group which is responsible for its inhibitory action towards protein tyrosine phosphatase 1B (PTP1B).