Learn More
Phenomenex Inc Claricep Flash Silica Column
Catalog No. 502412725 Shop All Phenomenex Products
Encompass_Preferred
Spherical 20-35m 100, 220g
Catalog No. 50-241-2725 Supplier Phenomenex Inc Supplier No. SS1302200
Spherical 20-35m 100, 220g