Learn More
Phenomenex Inc Flash Spherical Silica column
Catalog No. 502412718 Shop All Phenomenex Products
Encompass_Preferred
Flash Spherical Silica column Spherical; 20-35µm 60 A; 4g/column
Catalog No. 50-241-2718 Supplier Phenomenex Inc Supplier No. SS1300040S
Flash Spherical Silica column Spherical; 20-35µm 60 A; 4g/column