Learn More
Phenomenex Inc Flash Spherical C18
Catalog No. 502399400 Shop All Phenomenex Products
Encompass_Preferred
Flash Spherical C18 Claricep Flash columns, spherical C18 40-60µm, 100A, 120g/Cartridge
Catalog No. 50-239-9400 Supplier Phenomenex Inc Supplier No. SO2401200
Flash Spherical C18 Claricep Flash columns, spherical C18 40-60µm, 100A, 120g/Cartridge