Learn More
Phenomenex Inc Flash Spherical C18
Catalog No. 502399413 Shop All Phenomenex Products
Encompass_Preferred
Flash Spherical C18 Claricep Flash columns, spherical C18 40-60µm, 100A, 80g/Cartridge
Catalog No. 50-239-9413 Supplier Phenomenex Inc Supplier No. SO2400800
Flash Spherical C18 Claricep Flash columns, spherical C18 40-60µm, 100A, 80g/Cartridge