Learn More
Phenomenex Inc Claricep Screw-on Flash AQ C18 Column
Catalog No. 502412671 Shop All Phenomenex Products
Encompass_Preferred
Spherical, 20-35m, 100, 80g, S-Series, with Luer
Catalog No. 50-241-2671 Supplier Phenomenex Inc Supplier No. SSQ2300800
Spherical, 20-35m, 100, 80g, S-Series, with Luer