Learn More
Phenomenex Inc Claricep Screw-on Flash C18 Column
Catalog No. 502412648 Shop All Phenomenex Products
Encompass_Preferred
Spherical, 40-60m, 100, 20g, S-Series, with Luer
Catalog No. 50-241-2648 Supplier Phenomenex Inc Supplier No. SSO2400200
Spherical, 40-60m, 100, 20g, S-Series, with Luer