Learn More
Phenomenex Inc Claricep Screw-on Flash C18 Column
Catalog No. 502398541 Shop All Phenomenex Products
Encompass_Preferred
Claricep Screw-on Flash C18 Column Spherical; 40-60μm; 100A; 20g; C-Series
Catalog No. 50-239-8541 Supplier Phenomenex Inc Supplier No. CSO2400200
Claricep Screw-on Flash C18 Column Spherical; 40-60μm; 100A; 20g; C-Series