Learn More
Phenomenex Inc Claricep Screw-on Flash HILIC column
Catalog No. 502412657 Shop All Phenomenex Products
Encompass_Preferred
Spherical, 20-35µm, 100A, 20g, S-Series, with luer lock, 1/pk
Catalog No. 50-241-2657 Supplier Phenomenex Inc Supplier No. SSH2300200S
Spherical, 20-35µm, 100A, 20g, S-Series, with luer lock, 1/pk