Learn More
Phenomenex Inc Claricep Screw-on Flash HILIC Column
Catalog No. 502412840 Shop All Phenomenex Products
Encompass_Preferred
Spherical, 20-35m, 100, 20g, S-Series, with Luer
Catalog No. 50-241-2840 Supplier Phenomenex Inc Supplier No. SSH2300200
Spherical, 20-35m, 100, 20g, S-Series, with Luer