Learn More
Phenomenex Inc Claricep Screw-on Flash NH2 Column
Catalog No. 502412642 Shop All Phenomenex Products
Encompass_Preferred
Spherical, 40-60m, 100, 80g, S-Series, with Luer
Catalog No. 50-241-2642 Supplier Phenomenex Inc Supplier No. SSN2400800
Spherical, 40-60m, 100, 80g, S-Series, with Luer