Learn More
Azer Scientific Clarifying Differentiator, iCON™ Staining System
SDP
Catalog No. 21377142
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-142 4 x 1 gal.
21-377-141 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-377-142 Supplier Azer Scientific Supplier No. ES365514G
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to be used after hematoxylin staining, increases transparency, definition of hematoxylin and eosin staining.
  • Increases transparency and definition of hematoxylin and eosin staining
  • Clarifying differentiator has the additional benefits of removing nonspecific hematoxylin and slide adhesive staining
  • Recommended exposure time to the differentiator is thirty seconds to one minute
  • Can be used with any staining protocol to improve staining results
  • May be used on all automatic stainers as well as in manual staining
  • Two year shelf life
Specifications
Quantity 4 x 1 gal.