Learn More
MSC Betco Foaming Hand Sanitizer, Alcohol Free, 1000 mL, Clario C3 Touch Free
Catalog No. 502839224
Encompass
This Alcohol Free Foaming Hand Sanitizer uses 0.13% benzalkonium chloride for germ reduction. Its creamy, emollient-enriched lather prevents dryness and irritation, encouraging frequent use. Alcohol-free and in sealed packaging, it ensures purity and efficient product use in any environment.
  • Contains 0.13% benzalkonium chloride for germ reduction.
  • Creamy foaming lather enriched with emollients, gentle on skin and prevents dryness or irritation.
  • Does not contain alcohol, suitable for any environment.
  • Thick, rich foam encourages frequent use.
  • Use as needed between hand washes to help reduce bacteria on the skin.
  • Fresh neutral scent for pleasant hand sanitizing experience.
  • Sealed packaging keeps formula free of contaminants and provides 99% product evacuation.
Catalog No. 50-283-9224 Supplier MSC Supplier No. 73069791
This Alcohol Free Foaming Hand Sanitizer uses 0.13% benzalkonium chloride for germ reduction. Its creamy, emollient-enriched lather prevents dryness and irritation, encouraging frequent use. Alcohol-free and in sealed packaging, it ensures purity and efficient product use in any environment.
  • Contains 0.13% benzalkonium chloride for germ reduction.
  • Creamy foaming lather enriched with emollients, gentle on skin and prevents dryness or irritation.
  • Does not contain alcohol, suitable for any environment.
  • Thick, rich foam encourages frequent use.
  • Use as needed between hand washes to help reduce bacteria on the skin.
  • Fresh neutral scent for pleasant hand sanitizing experience.
  • Sealed packaging keeps formula free of contaminants and provides 99% product evacuation.