Learn More
MilliporeSigma Clarisolve 60 µm Pod Depth Filters, 0.33 m2 (3.55 ft2)
Supplier:  MilliporeSigma CS60HX03F1X
Encompass
Clarisolve 60 µm Pod Depth Filters, 0.33 m2 (3.55 ft2)
Catalog No. NC1714187