Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claritas PPT™ Grade Chromium, 10 μg/mL (10 ppm) For ICP/ICP-MS in 2% HNO3, SPEX CertiPrep™
SDP

Catalog No. p-7221239 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
30 mL
125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLCR21AM 30 mL
CLCR21AY 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CLCR21AM Supplier SPEX CertiPrep Supplier No. CLCR21AM
Add to Cart
Add to Cart

Single-element solutions at 10 ppm within 2% of uncertainty, made using ultra-high purity acids, the highest-grade starting materials and a high purity water

0

Specifications

Packaging Triple-leached LDPE bottle
CAS 7697-37-2, 7732-18-5, 7440-47-3
CAS Max % 0.02, 0.97999, 1e-05
Physical Form Liquid
Concentration 10 μg/mL
Technique ICP, ICP-MS
UN Number UN3264
Solubility Miscible with water
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Solvent or Matrix 2% HNO3
Quantity 30 mL
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.