Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claritas PPT™ Grade Iron, 10 μg/mL (10 ppm) For ICP/ICP-MS in 2% HNO3, SPEX CertiPrep™
Shop All Spex CertiPrep Products

Specifications
Specifications
| Packaging | Triple-leached LDPE bottle |
| CAS | 7697-37-2, 7732-18-5, 7439-89-6 |
| CAS Max % | 0.02, 0.97999, 1e-05 |
| Physical Form | Liquid |
| Concentration | 10 μg/mL |
| Technique | ICP, ICP-MS |
| UN Number | UN3264 |
| Solubility | Miscible with water |
| Solution Type | Standard Solution |
| Type | ICP/ICP-MS Standard |
| Show More |
Product intended for laboratory use only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?