Learn More
StatLab ClariTex Supa Mega Coverslips

Description
- All coverslips fit Supa Mega Microscope Slides and feature a uniform thickness and flatness
- Manufactured to DIN ISO 8255
- BX100
Specifications
Specifications
| Product Type | Premium Coverslip |
| Shape | Rectangle |
| Material | Borosilicate Glass D 263 M |
| Length (Metric) | 64 mm |
| Width (Metric) | 50 mm |
| Thickness (Metric) | 1.5 |
| Sterility | Non-sterile |
| Disposable | Single-use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.