Learn More
StatLab ClariTex Supa Mega Coverslips
SDP
Catalog No. 22000077
Change view
Click to view available options
Thickness (Metric):
1
1.5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (Metric)
22-000-077 1
22-000-076 1.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-077 Supplier StatLab Supplier No. SAG506402A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed with the specifications of Supa Mega Microscope Slides in mind
  • All coverslips fit Supa Mega Microscope Slides and feature a uniform thickness and flatness
  • Manufactured to DIN ISO 8255
  • BX100
Specifications
Product Type Premium Coverslip
Shape Rectangle
Material Borosilicate Glass D 263 M
Length (Metric) 64 mm
Width (Metric) 50 mm
Thickness (Metric) 1
Sterility Non-sterile
Disposable Single-use