Learn More
Apexbio Technology LLC Clarithromycin,100mg CAS# 81103-11-9
SDP
Supplier:  Apexbio Technology LLC A4322100
Encompass_Preferred
Clarithromycin, is a potent inhibitor of the cytochrome P450 isoenzyme CYP3A, might build the plasma concentration of statins metabolized by this pathway; therefore, increase the risk of interaction with statins in reference to pharmacokinetic studies. Among 28,484 medicines of clarithromycin, 2317 persons were co-presented to statins. Co-organization of CYP3A4 metabolized statins and clarithromycin was connected with a 2.11 fold expanded danger of death or hospitalization (95 % certainty interim [CI]: 1.79-2.48). Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0639