Learn More
Cayman Chemical Clarithromycin-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2699608-30-3, 1 mg
SDP
Catalog No. 502935282
Encompass_Preferred
An internal standard for the quantification of clarithromycin by GC- or LC-MS
Catalog No. 50-293-5282 Supplier Cayman Chemical Supplier No. 266781 MG
An internal standard for the quantification of clarithromycin by GC- or LC-MS